Learn More
Invitrogen™ PAX5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA138418
Description
This antibody is predicted to react with chicken, canine, mouse based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is tonsil tissue.
PAX5 is a member of the paired box (PAX) family of transcription factors. The PAX proteins are important regulators in early development and alterations in the expression of their genes are thought to contribute to neoplastic transformation. PAX5 is the B-cell lineage specific activator protein (BSAP) that is expressed at early, but not late stages, of B-cell differentiation. Its expression has also been detected in developing CNS and testis, therefore, PAX5 may not only play an important role in B-cell differentiation, but also in neural development and spermatogenesis. Mutations in the gene can result in leukemia, acute lymphoblastic 3.
Specifications
| PAX5 | |
| Polyclonal | |
| PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
| Q02548 | |
| PAX5 | |
| Synthetic peptide derived from C-terminus of human Pax-5 protein. | |
| 1 mL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| PAX5 | |
| ALL3; B-cell lineage specific activator; B-cell-specific transcription factor; BSAP; EBB-1; KLP; paired box 5; paired box gene 5; paired box gene 5 (B-cell lineage specific activator); paired box homeotic gene 5; paired box protein Pax-5; paired domain gene 5; Pax5; Pax-5; Pax5 variant 1; Pax5 variant 2; RGD1565236; transcription factor PAX 5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5079 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.