Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pax6 Mouse anti-Chicken, Human, Mouse, Rat, Zebrafish, Clone: AD1.5, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB5554
Description
Anti-Pax6 Antibody, clone AD1.5 is an antibody against Pax6 for use in immunohistochemistry and western blotting.
Specifications
Pax6 | |
Monoclonal | |
Unconjugated | |
NM_000280.2; NM_001604.3 | |
Mouse | |
Unpurified | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Immunohistochemistry, Western Blot | |
AD1.5 | |
Ascites fluid containing no preservatives. | |
PAX6, WAGR, Oculorhombin, D11S812E, AN2, MGDA, MGC17209, AN | |
Recombinant human Pax6. | |
100 μL | |
Chicken, Human, Mouse, Rat, Zebrafish | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction