Learn More
MilliporeSigma™ anti-Pax6, Clone: AD1.5, (Chemicon)
Supplier: MilliporeSigma™ MAB5554/DEL

Description
This gene encodes paired box gene 6, one of many human homologues of the Drosophila melanogaster gene prd. In addition to the hallmark feature of this gene family, a conserved paired box domain, the encoded protein also contains a homeo box domain. Both domains are known to bind DNA, and function as regulators of gene transcription. This gene is expressed in the developing nervous system, and in developing eyes. Mutations in this gene are known to cause aniridia as well as Peter's anomaly, both ocular diseases.
Immunohistochemistry, Western Blotting

Specifications
Pax6 | |
AD1.5 | |
Mouse | |
100 μL | |
Neuroscience | |
Chicken, Human, Mouse, Rat, Zebrafish | |
IgG1 |
Monoclonal | |
Unconjugated | |
Recombinant human Pax6. | |
RUO | |
Primary | |
Ascites |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.