Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paxillin Mouse, Unlabeled, Clone: 165, BD

Catalog No. BDB610620 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610620 150 μg
BDB610619 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610620 Supplier BD Biosciences Supplier No. 610620
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Paxillin, a focal adhesion protein, is a substrate for several tyrosine kinases such as src, FAK, and p120BRC/ABL. The tyrosine phosphorylation of paxillin is affected by conditions that change cell-cell adhesion. This is consisent with the possibility that paxillin is involved in the regulation of cell morphology. Additionally, because of its SH3 binding domain, paxillin associates tightly with FAK and Crk in an extracellular matrix-independent manner. Paxillin was initially detected in fibroblasts, and its phosphorylation may be important during neurite extension during differentiation.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Paxillin
Applications Western Blot
Classification Monoclonal
Clone 165
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.