Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paxillin Mouse anti-Human, Rat, Mouse, Bovine, Avian, Clone: 5H11, MilliporeSigma™

Catalog No. 05417MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-417-MI 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-417-MI Supplier MilliporeSigma Supplier No. 05417
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Paxillin Antibody, clone 5H11 is an antibody against Paxillin for use in immunocytochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Paxillin
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 5H11
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 in buffer containing 0.1MTris-glycine, pH 7.4, 0.15MNaCl, 0.05% sodium azide. Store at −20°C.
Gene Accession No. NM_002859
Gene Symbols PXN, Paxillin, paxillin, FLJ16691
Host Species Mouse
Immunogen GST fusion protein containing fulllength human paxillin. Clone 5H11.
Purification Method Protein G purfied
Quantity 250 μg
Primary or Secondary Primary
Target Species Avian, Bovine, Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.