Learn More
Description
Specifications
Specifications
| Antigen | Paxillin |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity-purified antibody is supplied in 100 #956;l phosphate-buffered saline, 50% glycerol, 1mg/mL BSA, and 0.05% sodium azide. |
| Gene Symbols | PXN; Paxillin.; paxillin; FLJ16691 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide (coupled to KLH) corresponding to amino acid residues around serine 178 of human paxillin. This human sequence is highly conserved in rat and mouse paxillin. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.