Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

propionyl Coenzyme A carboxylase, alpha polypeptide, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016873 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-873 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-873 Supplier Abnova Corporation Supplier No. H00005095B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human PCCA protein.

The protein encoded by this gene is the alpha subunit of the heterodimeric mitochondrial enzyme Propionyl-CoA carboxylase. PCCA encodes the biotin-binding region of this enzyme. Mutations in either PCCA or PCCB (encoding the beta subunit) lead to an enzyme deficiency resulting in propionic acidemia. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MLSAALRTLKHVLYYSRQCLMVSRNLGSVGYDPNEKTFDKILVANRGEIACRVIRTCKKMGIKTVAIHSDVDASSVHVKMADEAVCVGPAPTSKSYLNMDAIMEAIKKTRAQAVHPGYGFLSENKEFARCLAAEDVVFIGPDTHAIQAMGDKIESKLLAKKAEVNTIPGFDGVVKDAEEAVRIAREIGYPVMIKASAGGGGKGMRIAWDDEETRDGFRLSSQEAASSFGDDRLLIEKFIDNPRHIEIQVLGDKHGNALWLNERECSIQRRNQKVVEEAPSIFLDAETRRAMGEQAVALARAVKYSSAGTVEFLVDSKKNFYFLEMNTRLQVEHPVTECITGLDLVQEMIRVAKGYPLRHKQADIRINGWAVECRVYAEDPYKSFGLPSIGRLSQYQEPLHLPGVRVDSGIQPGSDISIYYDPMISKLITYGSDRTEALKRMADALDNYVIRGVTHNIALLREVIINSRFVKGDISTKFLSDVYPDGFKGHMLTKSEKNQLLAIASSLFVAFQLRAQHFQENSRMPVIKPDIANWELSVKLHDKVHTVVASNNGSVFSVEVDGSKLNVTSTWNLASPLLSVSVDGTQRTVQCLSREAGGNMSIQFLGTVYKVNILTRLAAELNKFMLEKVTEDTSSVLRSPMPGVVVAVSVKPGDAVAEGQEICVIEAMKMQNSMTAGKTGTVKSVHCQAGDTVGEGDLLVELE

Specifications

Antigen propionyl Coenzyme A carboxylase, alpha polypeptide
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human PCCA protein.
Formulation PBS with no preservative; pH 7.4
Gene PCCA
Gene Accession No. BC000140.2
Gene Symbols PCCA
Host Species Mouse
Immunogen PCCA (AAH00140.1, 1 a.a. ∼ 703 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5095
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.