Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

protocadherin 1, Mouse, Clone: 4H2, Abnova™

Catalog No. 89000865 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-865 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-865 Supplier Abnova Corporation Supplier No. H00005097M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant PCDH1.

This gene belongs to the protocadherin subfamily within the cadherin superfamily. The encoded protein is a membrane protein found at cell-cell boundaries. It is involved in neural cell adhesion, suggesting a possible role in neuronal development. The protein includes an extracelllular region, containing 7 cadherin-like domains, a transmembrane region and a C-terminal cytoplasmic region. Cells expressing the protein showed cell aggregation activity. Alternative splicing occurs in this gene. [provided by RefSeq

Sequence: YKVPEEQPPNTLIGSLAADYGFPDVGHLYKLEVGAPYLRVDGKTGDIFTTETSIDREGLRECQNQLPGDPCILEFEVSITDLVQNGSPRLLEGQIEVQDINDNTPNFA

Specifications

Antigen protocadherin 1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4H2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant PCDH1.
Formulation PBS with no preservative; pH 7.4
Gene PCDH1
Gene Accession No. NM_002587
Gene Alias FLJ53887/MGC45991/PC42/PCDH42
Gene Symbols PCDH1
Host Species Mouse
Immunogen PCDH1 (NP_002578, 62 a.a. ∼ 169 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 5097
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.