Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

protocadherin 21, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89008426 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-008-426 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-008-426 Supplier Abnova Corporation Supplier No. H00092211B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human PCDH21 protein.

This gene is a member of the cadherin superfamily of calcium-dependent cell-cell adhesion molecules. The encoded protein has a signal peptide, six cadherin repeat domains and a unique cytoplasmic region. This nonclassical cadherin appears to be exclusively expressed in the mitral and tufted cells in the main and accessory olfactory bulbs of the brain, suggesting a possible role in the formation and maintenance of neuronal networks. [provided by RefSeq

Sequence: MRRCRWAALALGLLRLCLAQANFAPHFFDNGVGSTNGNMALFSLPEDTPVGSHVYTLNGTDPEGDPISYHISFDPSTRSVFSVDPTFGNITLVEELDREREDEIEAIISISDGLNLVAEKVVILVTDANDEAPRFIQEPYVALVPEDIPAGSIIFKVHAVDRDTGSGGSVTYFLQNLHSPFAVDRHSGVLRLQAGATLDYERSRTHYITVVAKDGGGRLHGADVVFSATTTVTVNVEDVQDMAPVFVGTPYYGYVYEDTLPGSEVLKVVAMDGDRGKPNRILYSLVNGNDGAFEINETSGAISITQSPAQLQREVYELHVQVTEMSPAGSPAAQATVPVTIRIVDLNNHPPTFYGESGPQNRFELSMNEHPPQGEILRGLKITVNDSDQGANAKFNLQLVGPRGIFRVVPQTVLNEAQVTIIVENSAAIDFEKSKVLTFKLLAVEVNTPEKFSSTADVVIQLLDTNDNVPKFDSLYYVARIPENAPGGSSVVAVTAVDPDTGPWGEVKYSTYGTGADLFLIHPSTGLIYTQPWASLDAEATARYNFYVKAEDMEGKYSVAEVFITLLDVNDHPPQFGKSVQKKTMVLGTPVKIEAIDEDAEEPNNLVDYSITHAEPANVFDINSHTGEIWLKNSIRSLDALHNITPGRDCLWSLEVQAKDRGSPSFSTTALLKIDITDAEVRRLRYMKNSNFPGTTKSVRKPKFKPKKPHSSQGLFLHPHCEIALFNLSNVNLYSRVFQGAAQAS

Specifications

Antigen protocadherin 21
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human PCDH21 protein.
Formulation No additive
Gene PCDH21
Gene Accession No. BC038799.1
Gene Alias DKFZp434A132/KIAA1775/PRCAD
Gene Symbols PCDH21
Host Species Mouse
Immunogen PCDH21 (AAH38799.1, 1 a.a. ∼ 745 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cell Adhesion
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 92211
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.