Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

protocadherin 7, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004228 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-228 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-228 Supplier Abnova Corporation Supplier No. H00005099A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PCDH7.

This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The gene encodes a protein with an extracellular domain containing 7 cadherin repeats. The gene product is an integral membrane protein that is thought to function in cell-cell recognition and adhesion. Alternative splicing yields isoforms with unique cytoplasmic tails. [provided by RefSeq

Sequence: KQLLRYRLAEEGPADVRIGNVASDLGIVTGSGEVTFSLESGSEYLKIDNLTGELSTSERRIDREKLPQCQMIFDENECFLDFEVSVIGPSQSWV

Specifications

Antigen protocadherin 7
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PCDH7.
Formulation 50% glycerol
Gene PCDH7
Gene Accession No. NM_002589
Gene Alias BH-Pcdh/BHPCDH
Gene Symbols PCDH7
Host Species Mouse
Immunogen PCDH7 (NP_002580, 31 a.a. ∼ 124 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cell Adhesion
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 5099
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.