Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

protocadherin alpha 5, Mouse, Clone: 1E8, Abnova™

Catalog No. 89001543 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-543 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-543 Supplier Abnova Corporation Supplier No. H00056143M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant PCDHA5.

This gene is a member of the protocadherin alpha gene cluster, one of three related gene clusters tandemly linked on chromosome five that demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The alpha gene cluster is composed of 15 cadherin superfamily genes related to the mouse CNR genes and consists of 13 highly similar and 2 more distantly related coding sequences. The tandem array of 15 N-terminal exons, or variable exons, are followed by downstream C-terminal exons, or constant exons, which are shared by all genes in the cluster. The large, uninterrupted N-terminal exons each encode six cadherin ectodomains while the C-terminal exons encode the cytoplasmic domain. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins that most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been observed and additional variants have been suggested but their full-length nature has yet to be determined. [provided by RefSeq

Sequence: TNEEETNFLELVLRKSLDREETQEHRLLVIATDGGKPELTGTVQLLINVLDANDNAPEFDKSIYNVRLLENAPSGTLVIKLNASDADEGINKEIVYFFSNLVLDDVK

Specifications

Antigen protocadherin alpha 5
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1E8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant PCDHA5.
Formulation ascites with no preservative
Gene PCDHA5
Gene Accession No. NM_018908
Gene Alias CNR6/CNRN6/CNRS6/CRNR6/PCDH-ALPHA5
Gene Symbols PCDHA5
Host Species Mouse
Immunogen PCDHA5 (NP_061731, 183 a.a. ∼ 289 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Research Discipline Cell Adhesion
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 56143
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.