Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

protocadherin beta 16, Mouse, Clone: 3F12-2A6, Abnova™

Catalog No. 89001328 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-328 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-328 Supplier Abnova Corporation Supplier No. H00057717M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant PCDHB16.

This gene is a member of the protocadherin beta gene cluster, one of three related gene clusters tandemly linked on chromosome five. The gene clusters demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The beta cluster contains 16 genes and 3 pseudogenes, each encoding 6 extracellular cadherin domains and a cytoplasmic tail that deviates from others in the cadherin superfamily. The extracellular domains interact in a homophilic manner to specify differential cell-cell connections. Unlike the alpha and gamma clusters, the transcripts from these genes are made up of only one large exon, not sharing common 3' exons as expected. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins. Their specific functions are unknown but they most likely play a critical role in the establishment and function of specific cell-cell neural connections. [provided by RefSeq]

Sequence: MEIGWMHNRRQRQVLVFFVLLSLSGAGAELGSYSVVEETERGSFVANLGKDLGLGLTEMSTRKARIISQGNKQHLQLKAQTGDLLINEKLDREELCGPTEPCILHFQVLMENPLEIFQAELRVIDINDHSPMFTEKEMILKIPENSPLGTEFPLNHALDLDVGSNNVQNYKISPSSHFRVLIHEFRDGRKYPELVLDKELDREEEPQLRLTLTALDGGSPPRSGTAQVRIEVVDINDNAPEFEQPIYKVQIPENSPLGSLVATVSARDLDGGANGKISYTLFQPSEDISKTLEVNPMTGEVRLRKQVDFEMVTSYEVRIKATDGGGLSGKCTLLLQVVDVNDNPPQVTMSALTSPIAENSPEIVVAVFSVSDPDSGNNGKTISSIQEDLPFLLKPSVKNFYTLVTERALDREARAEYNITLTVTDMGTPRLKTEHNITVQISDVNDNAPTFTQTSYTLFVRENNSPALHIGSVSATDRDSGTNAQVTYSLLPPQDPHLPLASLVSINADNGHLFALRSLDYEALQAFEFRVGATDRGSPALSREALVRVLVLDANDNSPLVLYPLQNGSAPCTELVPRAAEPGYLVTKVVAVDGDSGQNAWLSYQLLKATEPGLFGVWAHNGEVRTARLLSERDAAKQRLVVLVKDNGEPPRSATATLHVLLVDGFSQPFLPLPEAAPGQTQANSLTVYLVVALASVSSLFLFSVLLFVAVRLCRRSRAASVGRCSMPEGPFPGRLVDVSGTGTLSQSYQYEVCLTGGSETSEFKFLKPIIPNFSP

Specifications

Antigen protocadherin beta 16
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3F12-2A6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant PCDHB16.
Formulation PBS with no preservative; pH 7.4
Gene PCDHB16
Gene Accession No. BC036062
Gene Alias KIAA1621/ME1/PCDH-BETA16/PCDH3X/PCDHB8a
Gene Symbols PCDHB16
Host Species Mouse
Immunogen PCDHB16 (AAH36062.1, 1 a.a. ∼ 776 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Adhesion
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 57717
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.