Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCMT-I/II Mouse, Unlabeled, Clone: 4, BD

Catalog No. BDB610772 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610772 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610772 Supplier BD Biosciences Supplier No. 610772

Mouse Monoclonal Antibody

PCMT-II (Protein Carboxyl Methyl Transferase), or L-isoaspartate (D-aspartate) O-methyltransferase, is a cytosolic enzyme that is expressed in all tissues as well as in bacteria. Interestingly, PCMT-II is highly expressed in the brain. It recognizes damaged proteins that result from deamidation, isomerization, and racemization reactions. These proteins contain L-isoaspartyl and D-aspartyl residues and can have abnormal configurations that result in anomalous or loss of function. PCMT-II catalyzes the first step of the conversion of these unusual residues to normal L-aspartyl residues. This mechanism prevents the accumulation of aberrant proteins in aging cells. Pcmt-1 knock-out mice display growth retardation and a seizure disorder that results in death at an average age of 42 days. Thus, PCMT-II is necessary for growth and nervous system function. The immunogen region from human PCMT-II used to generate this antibody has been reported to be identical to the region in PCMT-I.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen PCMT-I/II
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 4
Concentration 250μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human PCMT-II aa. 10-211
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Bovine, Canine, Human
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.