Learn More
PCNA Mouse anti-Human, Biotin, Clone: PC10, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 555567
Description
Glycophorin A is a sialoglycoprotein present on human red blood cells (RBC) and erythroid precursor cells. This antibody recognizes human RBCs and erythroid precursors and is useful in erythroid cell development studies. Mature, non-nucleated red blood cells are characteristically glycophorin A positive.
Intracellular Staining

Specifications
PCNA | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Not Reported | |
0.1 mg | |
Cell Biology | |
Human | |
IgG2a κ |
Flow Cytometry | |
PC10 | |
Biotin | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.