Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCNA Mouse, Unlabeled, Clone: 24, BD

Catalog No. BDB610664 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610664 50 μg
BDB610665 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610664 Supplier BD Biosciences Supplier No. 610664
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Progression of the mammalian cell cycle is regulated in two different ways: 1) phosphorylation and dephosphorylation of key proteins, 2) synthesis and degradation of regulatory factors. The Proliferating Cell Nuclear Antigen (PCNA) was initially identified as a nuclear antigen in proliferating cells and was subsequently described as a subunit for DNA polymerase δ. Human PCNA is 262 amino acids with an apparent molecular weight of 36kDa. PCNA protein levels peak during the S-phase of the cell cycle, at which time it forms a complex with the p21 inhibitor. PCNA is almost undetectable in other phases of the cycle. Because of its unique expression, PCNA has been extensively used in studies associating the prognosis of tumor progression and neoplastic proliferation.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen PCNA
Applications Western Blot
Classification Monoclonal
Clone 24
Concentration 250μg/mL
Conjugate Unconjugated
Description Proliferating Cell Nuclear Antigen
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.