Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCNA Mouse anti-Vertebrates, Clone: PC10, MilliporeSigma™

Catalog No. 05347MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-347-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-347-MI Supplier MilliporeSigma Supplier No. 05347
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect PCNA with Anti-PCNA Antibody, clone PC10 (Mouse Monoclonal Antibody), that has been shown to work in western blot, immunocytochemistry and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCNA
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone PC10
Conjugate Unconjugated
Formulation Protein A purified mouse immunoglobulin in PBS containing 1mg/mL BSA and 10mM Sodium Azide.
Gene Accession No. NM_002592
Gene Symbols PCNA, Cyclin, cyclin, MGC8367
Host Species Mouse
Immunogen Recombinant rat PCNA
Purification Method Concentrated culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Target Species Vertebrates
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.