Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCNA Mouse anti-Invertebrates, Vertebrates, Clone: PC10, MilliporeSigma™

Catalog No. MAB424MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB424MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB424MI Supplier MilliporeSigma Supplier No. MAB424
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-PCNA Antibody, clone PC10 (mouse monoclonal antibody) validated in ELISA, flow cytometry,immunohistochemistry (paraffin), immunoprecipitation, western blotting to detect PCNA also known as Proliferating Cell Nuclear Antigen, DNA Polymerase delta Processivity Factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCNA
Applications ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone PC10
Conjugate Unconjugated
Formulation Protein A Purified mouse immunoglobulin in PBS with 0.1% sodium azide as a preservative.
Gene Accession No. NM_002592.2; NM_182649.1
Gene Symbols PCNA, Cyclin, cyclin, MGC8367
Host Species Mouse
Immunogen Rat PCNA made in the protein A vector pR1T2T.
Purification Method Protein A purfied
Quantity 500 μg
Primary or Secondary Primary
Target Species Invertebrate, Vertebrates
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.