Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCNA Mouse anti-Human, Clone: PC10, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABE288
Description
Use Anti-PCNA Antibody, clone PC10 (Mouse Monoclonal Antibody) validated in western blot, immunohistochemistry, immunofluorescence to detect PCNA also known as Proliferating cell nuclear antigen, PCNA, Cyclin.
Specifications
PCNA | |
Monoclonal | |
Unconjugated | |
NP_002583 | |
Mouse | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG2a κ |
Immunofluorescence, Immunohistochemistry, Western Blot | |
PC10 | |
Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
PCNA | |
Recombinant protein corresponding to human PCNA. | |
200 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction