Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PCNA Clone: PC10,

Catalog No. NA03200UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
NA03200UG 200 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NA03200UG Supplier MilliporeSigma™ Supplier No. NA03200UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects PCNA Clone: PC10 in Human, Mouse, Rat, Yeast, Zebrafish samples, and it is validated for Immunofluorescence, Immunohistochemistry (Paraffin)

The proliferating cell nuclear antigen (PCNA) is a 37kDa molecular weight protein also known as cyclin. The protein has also been identified as the polymerase δ accessory protein and is detected in a cell cycle dependent manner. In early S phase, PCNA has a very granular distribution and is absent from the nucleoli. At late S phase, PCNA is prominent in the nucleoli. In cells fixed with organic solvents, the PCNA is seen to be strongly associated in the nuclear regions where DNA synthesis is occurring, whereas in cells fixed with aldehydes, the staining is more diffuse but intense and occurs throughout the cell cycle. This is due to the presence of two basic forms of the PCNA protein: a soluble form which is sensitive to organic fixation and not involved in replication and a second form which is insoluble and associated with on-going DNA synthesis. PCNA is a very conserved protein present not only in mammals but also in plant cells. Patients suffering from SLE have been shown to have auto antibodies against PCNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCNA
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PC10
Conjugate Unconjugated
Formulation In 50mM sodium phosphate buffer, 0.2% gelatin, pH 7.5.
Host Species Mouse
Immunogen recombinant PCNA
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽37kDa PCNA protein in HeLa cells as well as in tonsil or normal colon tissue.
Target Species Human, Mouse, Rat, Yeast, Zebrafish
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.