Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE4D Rabbit anti-Rat, Human, Chimpanzee, Polyclonal, MilliporeSigma™

Catalog No. ABS22MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS22MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS22MI Supplier MilliporeSigma Supplier No. ABS22
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-PDE4D Antibody is an antibody against PDE4D for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDE4D
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols PDE4DN2; DPDE3; PDE43; PDE4D; STRK1; HSPDE4D
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to the C-terminus of human PDE4D.
Purification Method Protein A
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_001098101
Target Species Chimpanzee, Human, Rat
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.