Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDE9A Rabbit anti-Human, Mouse, Rhesus Macaque, Rat, Chimpanzee, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABN32
Description
Detect PDE9A using this Anti-PDE9A Antibody validated for use in Western Blotting, immunohistochemistry (paraffin).
Specifications
PDE9A | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Protein A purified | |
Primary | |
Chimpanzee, Human, Mouse, Rat, Rhesus Monkey | |
Purified |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
PDE9A | |
KLH-conjugated linear peptide corresponding to human PDE9A. | |
100 μg | |
NP_002597 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction