Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGF-AA Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 071436MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-143-6MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-143-6MI Supplier MilliporeSigma Supplier No. 071436
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect PDGF-AA using this Anti-PDGF-AA Antibody validated for use in immunohistochemistry (paraffin), ELISA, western blot, NEUT.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDGF-AA
Applications ELISA, Immunohistochemistry (Paraffin), Neutralization, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG lyophilized from PBS, pH 7.2.
Gene Symbols PDGFA; PDGF-1; PDGF-A; PDGF1
Host Species Rabbit
Immunogen Highly pure (>98%) recombinant human PDGF-AA.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_002607.4
Target Species Human, Mouse
Content And Storage Lyophilized: −20°C for up to 12 months. Reconstituted: 2-4°C in undiluted aliquots for up to 6 weeks
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.