Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGF-BB Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 071437MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-143-7MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-143-7MI Supplier MilliporeSigma Supplier No. 071437
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-PDGF-BB Antibody detects level of PDGF-BB and has been published and validated for use in western blot, ELISA, NEUT.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDGF-BB
Applications ELISA, Neutralization, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG lyophilized from 0.5X PBS, pH 7.4. Reconstitute with sterile distilled water.
Gene Symbols PDGFB; FLJ12858; PDGF2; c-sis; PDGF-2; SSV; SIS; Becaplermin
Host Species Rabbit
Immunogen Highly purified (>98%) recombinant human PDGF-BB
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_002608
Target Species Human
Content And Storage Lyophilized: 2-8°C for up to 12 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Avoid repeated freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.