Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGFRΒ Rabbit anti-Mouse, Human, Clone: 4A56, MilliporeSigma™

Catalog No. 04825MI Shop All MilliporeSigma Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-825-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. 04-825-MI Supplier MilliporeSigma Supplier No. 04825
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

This Anti-PDGFRβ Antibody, clone 4A56 is validated for use in immunoprecipitation, western blotting for the detection of PDGFRβ.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDGFRβ
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4A56
Conjugate Unconjugated
Formulation Cultured supernantant in 0.05% sodium azide
Gene Accession No. NM_002608
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 710-718 of human PDGF Receptor β. An N-terminus cysteine was added to facilitate conjugation. This sequence corresponds to amino acids 709-717 of mouse PDGF Receptor β.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) PDGF-R-beta, CD140B, JTK12, PDGFR, CD140b, PDGFR1, PDGFRB
Target Species Human, Mouse
Content And Storage −20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.