Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGFR Mouse anti-Human, Mouse, Rat, Clone: CH-3, MilliporeSigma™

Catalog No. 051135MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-113-5MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-113-5MI Supplier MilliporeSigma Supplier No. 051135
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-PDGFR Antibody, clone CH-3 is validated for use in western blot, ELISA, immunohistochemistry (paraffin) for the detection of PDGFR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDGFR
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CH-3
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 in buffer containing 20mM sodium phosphate, 250mM NaCl, 0.1% NaN3, pH. 7.6
Host Species Mouse
Immunogen Recombinant Platelet-Derived Growth Factor Receptor
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.