Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGFRα Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 07276MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-276-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-276-MI Supplier MilliporeSigma Supplier No. 07276
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect PDGFRα using this Anti-PDGFRα Antibody validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDGFRα
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Protein A Purified immunoglobulin in 0.14M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide as a preservative.
Gene Symbols PDGFRA; CD140a; PDGF-R-alpha; MGC74795; PDGFR2; Rhe-PDGFRA; CD140A
Host Species Rabbit
Immunogen peptide corresponding to amino acids 1035-1053 (C-GKRNRHSSQTSEESAIETG) of human PDGF Receptor α
Purification Method Affinity Purified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_011058
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months Avoid repeated freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.