Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDH Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526656
Description
This antibody is predicted to react with bovine, mouse, porcine, non-human primate and rat based on sequence homology.
The pyruvate dehydrogenase (PDH) complex is a nuclear-encoded mitochondrial multienzyme complex that catalyzes the overall conversion of pyruvate to acetyl-CoA and CO(2), and provides the primary link between glycolysis and the tricarboxylic acid (TCA) cycle. The PDH complex is composed of multiple copies of three enzymatic components: pyruvate dehydrogenase (E1), dihydrolipoamide acetyltransferase (E2) and lipoamide dehydrogenase (E3). The E1 enzyme is a heterotetramer of two alpha and two beta subunits. This gene encodes the E1 alpha 1 subunit containing the E1 active site, and plays a key role in the function of the PDH complex. Mutations in this gene are associated with pyruvate dehydrogenase E1-alpha deficiency and X-linked Leigh syndrome.
Specifications
PDH | |
Polyclonal | |
PDHA1 | |
PDHA, PDHCE1A, PHE1A, PDHE1-A type I, pyruvate dehydrogenase E1 component subunit alpha, somatic form, mitochondrial, pyruvate dehydrogenase complex, E1-alpha polypeptide 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5160 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
P08559 | |
PDHA1 | |
KLH conjugated synthetic peptide between 226-255 amino acids from the central region of human PDHA1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction