Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDK Mouse anti-Human, Rat, Clone: 3H1.2, MilliporeSigma™

Catalog No. 051912MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-191-2MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-191-2MI Supplier MilliporeSigma Supplier No. 051912

Mouse Monoclonal Antibody

This PDK antibody is validated for use in western blotting and immunocytochemistry for the detection of the PDK protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDK
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 3H1.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_002601
Gene Symbols PDK1, PDK
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to human PDK.
Purification Method Ion exchange
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.