Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pyruvate dehydrogenase kinase, isozyme 1, Mouse, Clone: 3B7, Abnova™

Catalog No. 89000871 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-871 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-871 Supplier Abnova Corporation Supplier No. H00005163M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant PDK1.

Pyruvate dehydrogenase (PDH) is a mitochondrial multienzyme complex that catalyzes the oxidative decarboxylation of pyruvate and is one of the major enzymes responsible for the regulation of homeostasis of carbohydrate fuels in mammals. The enzymatic activity is regulated by a phosphorylation/dephosphorylation cycle. Phosphorylation of PDH by a specific pyruvate dehydrogenase kinase (PDK) results in inactivation. [provided by RefSeq

Sequence: GGKGKGSPSHRKHIGSINPNCNVLEVIKDGYENARRLCDLYYINSPELELEELNAKSPGQPIQVVYVPSHLYHMVFELFKNAMRATMEHHANRGVYPPIQ

Specifications

Antigen pyruvate dehydrogenase kinase, isozyme 1
Applications ELISA
Classification Monoclonal
Clone 3B7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant PDK1.
Formulation PBS with no preservative; pH 7.4
Gene PDK1
Gene Accession No. BC039158
Gene Symbols PDK1
Host Species Mouse
Immunogen PDK1 (AAH39158, 203 a.a. ∼ 302 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Kinases
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 5163
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.