Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PDK2 Clone: 2G1,

Catalog No. ST1643100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ST1643100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ST1643100UG Supplier MilliporeSigma™ Supplier No. ST1643100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects PDK2 Clone: 2G1 in Human samples, and it is validated for ELISA, Immunohistochemistry (Paraffin), Immuno Blotting

The PDK2 protein inhibits the mitochondrial pyruvate dehydrogenase complex by phosphorylation of the E1 α subunit, thus contributing to the regulation of glucose metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDK2
Applications ELISA, Immunoblot, Immunoblot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2G1
Conjugate Unconjugated
Formulation In PBS, pH 7.2.
Host Species Mouse
Immunogen A recombinant polypeptide corresponding to amino acids of 187-276 human PDK2, expressed as a GST fusion protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽40kDa PDK2 protein in U-2 OS cells.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.