Learn More
PDK4, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human PDK4 peptide using ARM Technology.
Supplier: Abnova Corporation H00005166K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene is a member of the PDK/BCKDK protein kinase family and encodes a mitochondrial protein with a histidine kinase domain. This protein is located in the matrix of the mitrochondria and inhibits the pyruvate dehydrogenase complex by phosphorylating one of its subunits, thereby contributing to the regulation of glucose metabolism. Expression of this gene is regulated by glucocorticoids, retinoic acid and insulin. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
PDK4 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
PDK4 | |
PDK4 | |
A synthetic peptide of human PDK4 | |
RUO | |
5166 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
pyruvate dehydrogenase kinase, isozyme 4 | |
PDK4 | |
FLJ40832 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.