Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDPK1 Mouse, Unlabeled, Clone: 5, BD

Catalog No. BDB611070 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611070 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611070 Supplier BD Biosciences Supplier No. 611070

Mouse Monoclonal Antibody

Protein Kinase B (PKB) is downstream of phosphoinositide 3-kinase (PI3-Kinase) that functions in a critical signaling pathway for receptor-dependent cell activation. PKB activation requires the binding of lipid metabolites phosphatidylinositol 3,4,5-trisphosphate and phosphatidylinositol 3,4-bisphosphate, phospholipid products of PI3-Kinase. These lipids bind to PKB and induce a conformational change in the enzyme, resulting in its translocation to the plasma membrane. Following cell activation, PKB is phosphorylated on Thr-308 by another kinase designated as 3-Phosphoinositide-Dependent Protein Kinase 1 (PDPK1) or also known as PKB Kinase. This kinase probably represents a new family of phospholipid-dependent activities with molecular weights that range from 31 to 220kDa. PDPK1 consists of 559 amino acids with an N-terminal catalytic domain and a pleckstrin-homology (PH) domain located in the C-terminal region. Like PKB, PDPK1 requires the presence of phosphatidylinositol 3,4,5-trisphosphate for maximal activity. Thus, PDPK1 acts upstream of PKB and may control signals important for proliferation, apoptosis, and metabolism.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen PDPK1
Applications Western Blot
Classification Monoclonal
Clone 5
Concentration 250μg/mL
Conjugate Unconjugated
Description PKB Kinase, Akt Kinase, PDK1
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat PDPK1 aa. 439-555
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.