Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDPK1 (pS241) Mouse, PE, Clone: J66-653.44.17, BD

Catalog No. BDB560092 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560092 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560092 Supplier BD Biosciences Supplier No. 560092
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The serine/threonine kinase 3- Phosphoinositide- Dependent Protein Kinase-1 (PDPK1, also known as PDK1) contributes to the activation of many important kinases in the insulin and IGF-1 signaling pathways. It acts downstream of phosphatidylinositol 3-kinase (PI3-kinase) to phosphorylate residues in the activation loops of many cellular kinases, including protein kinase B (PKB/Akt), PKC isoforms, p70 S6 kinase, and PDPK1 itself. The autophosphorylation of PDPK1 at serine 241 (S241) has recently been suggested to play a role in the regulation of PDPK1. It has been proposed that PDPK1 activity plays a key role in the regulation of various cellular events such as cell proliferation, differentiation, and apoptosis.

The J666-653.44.17 monoclonal antibody recognizes the phosphorylated S241 in the activation loop of human PDPK1. The orthologous phosphorylation site in mouse and rat PDPK1 is S244.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PDPK1 (pS241)
Applications Flow Cytometry
Classification Monoclonal
Clone J66-653.44.17
Conjugate PE
Description PDK1 (pS241), PKB Kinase (pS241)
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human PDPK1 Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.