Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDX-1 Mouse anti-Human, Clone: 6F6.1, MilliporeSigma™

Catalog No. MAB4425MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4425MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4425MI Supplier MilliporeSigma Supplier No. MAB4425

Mouse Monoclonal Antibody

Anti-PDX-1 Antibody, clone 6F6.1 is a mouse monoclonal antibody for detection of PDX-1, also known as Pancreas Duodenum Homeobox-1 and insulin promoter factor 1 homeodomain transcription factor, and has been validated in Immunohistochemistry and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDX-1
Applications Western Blot
Classification Monoclonal
Clone 6F6.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_000200
Gene Symbols PDX1, IPF1, IDX-1, GSF, IUF1, IPF-1, STF-1, MODY4, PDX-1, IUF-1
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human PDX-1.
Purification Method Protein G Purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.