Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDX-1 Mouse, PerCP-Cy5.5, Clone: 658A5, BD

Catalog No. BDB563436 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563436 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563436 Supplier BD Biosciences Supplier No. 563436
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 658A5 monoclonal antibody binds PDX-1 or Pancreas/Duodenum Homeobox Protein-1 encoded by the PDX1 or IPF1 (Insulin Promoter Factor 1) gene. PDX-1 is a key regulator of pancreatic development and adult beta-cell function. Loss of PDX1 gene function in mice and humans results in pancreatic agenesis. PDX-1 is also involved in endocrine precursor cell development by binding the DNA-binding transcription factor Neurogenin-3. PDX-1-positive cells have been obtained by the directed differentiation of human embryonic stem cells through a definitive endoderm lineage.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PDX-1
Applications Flow Cytometry
Classification Monoclonal
Clone 658A5
Conjugate PerCP-Cyanine5.5
Description PDX1, GSF, IPF-1, IPF1, IUF-1, IDX-1, STF-1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human PDX-1 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.