Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pyridoxal (pyridoxine, vitamin B6) kinase (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89004397 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-397 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-397 Supplier Abnova Corporation Supplier No. H00008566A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PDXK.

The protein encoded by this gene phosphorylates vitamin B6, a step required for the conversion of vitamin B6 to pyridoxal-5-phosphate, an important cofactor in intermediary metabolism. The encoded protein is cytoplasmic and probably acts as a homodimer. Alternatively spliced transcript variants have been described, but their biological validity has not been determined. [provided by RefSeq

Sequence: HWKGQVLNSDELQELYEGLRLNNMNKYDYVLTGYTRDKSFLAMVVDIVQELKQQNPRLVYVCDPVLGDKWDGEGSMYVPEDLLPVYKEKVVPLADIITPNQFEAELLSGR

Specifications

Antigen pyridoxal (pyridoxine, vitamin B6) kinase
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PDXK.
Formulation 50% glycerol
Gene PDXK
Gene Accession No. NM_003681
Gene Alias C21orf124/C21orf97/DKFZp566A071/FLJ31940/FLJ37311/MGC15873/MGC31754/MGC52346/PKH/PNK/PRED79
Gene Symbols PDXK
Host Species Mouse
Immunogen PDXK (NP_003672, 51 a.a. ∼ 160 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 8566
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.