Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PDZK1, Clone: 1E9, Abnova

Catalog No. 89000875 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-875 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-875 Supplier Abnova Corporation Supplier No. H00005174M07

Mouse Monoclonal

ELISA, Immunohistochemistry (Paraffin), Sandwich ELISA, Western Blotting

Specifications

Antigen PDZ domain containing 1
Classification Monoclonal
Clone 1E9
Conjugate Unconjugated
Host Species Mouse
Immunogen PDZK1 (AAH06496, 1 a.a. ∽520 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Target Species Human
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.