Learn More
Description
Specifications
Specifications
| Antigen | PEAK1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified in PBS with 0.05% NaN3. |
| Gene Symbols | EC 2.7.10.2; FLJ21140; FLJ34483 ; KIAA2002 |
| Host Species | Rabbit |
| Immunogen | Recombinant Protein, aa: 1511-1660. Homology: 100% human and chimp; 96% pig and horse; 90% mouse; 88% rat |
| Purification Method | Affinity Purified |
| Quantity | 200 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.