Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-PECAM-1 (CD31), Clone: 2H8
Supplier: MilliporeSigma™ MAB1398Z/DEL

Description
Flow Cytometry

Specifications
PECAM-1 (CD31) | |
Unconjugated | |
100 μg | |
Cell Structure | |
Mouse | |
IgG |
Monoclonal | |
Hamster | |
RUO | |
Primary | |
Semi-purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction