Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-PECAM-1 (CD31), Clone: 390, ™ (Chemicon™)
Supplier: MilliporeSigma™ CBL1337/DEL
Description
Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Specifications
| PECAM-1 (CD31) | |
| Unconjugated | |
| Mouse leukocyte cell line 32D | |
| RUO | |
| Primary | |
| Semi-purified |
| Monoclonal | |
| Rat | |
| 500 μg | |
| Cell Structure | |
| Mouse | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction