Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PECAM-1 Armenian Hamster anti-Mouse, Canine, Clone: 2H8, MilliporeSigma™

Catalog No. MAB1398ZMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1398ZMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1398ZMI Supplier MilliporeSigma Supplier No. MAB1398Z
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

Anti-PECAM-1 Antibody, clone 2H8, Azide Free detects level of PECAM-1 and has been published and validated for use in Elecron Microscopy, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, and Neutralization Assay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PECAM-1
Applications Flow Cytometry
Classification Monoclonal
Clone 2H8
Conjugate Unconjugated
Formulation Protein A Purified mouse immunoglobulin in 20mM sodium phosphate, 250mM NaCl, pH. 7.6, containing no preservatives.
Gene Symbols ENSG00000188536, HBA2, Alpha-globin
Host Species Armenian Hamster
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.