Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PECAM-1 Rat anti-Mouse, Clone: 390, MilliporeSigma™

Catalog No. CBL1337MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL1337MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL1337MI Supplier MilliporeSigma Supplier No. CBL1337
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-PECAM-1 Antibody, clone 390 is an antibody against PECAM-1 for use in flow cytometry, immunohistochemistry and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PECAM-1
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 390
Conjugate Unconjugated
Formulation Protein A Purified mouse immunoglobulin in 100mM borate buffered saline, pH 8.0, containing no preservative.
Gene Accession No. NM_000442.3
Gene Symbols PECAM1, EndoCAM, GPIIA' , PECAM-1, CD31, CD31/EndoCAM
Host Species Rat
Immunogen Mouse leukocyte cell line 32D
Purification Method Protein A purfied
Quantity 500 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.