Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PECAM-1 Mouse anti-Human, Clone: P2B1, MilliporeSigma™

Catalog No. MAB2148CMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2148CMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2148CMI Supplier MilliporeSigma Supplier No. MAB2148C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-PECAM-1 Antibody, clone P2B1 (Ascites Free) is validated for use in flow cytometry, immunocytochemistry, immunoprecipitation, immunohistochemistry, ELISA and immunofluorescence for the detection of PECAM-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PECAM-1
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone P2B1
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_000433
Gene Symbols PECAM1
Host Species Mouse
Immunogen Whole cells expressing human PECAM-1.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.