Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PECAM-1 Mouse anti-Rat, Clone: TLD-3A12, MilliporeSigma™

Catalog No. MAB1393ZMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1393ZMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1393ZMI Supplier MilliporeSigma Supplier No. MAB1393Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-PECAM-1 Antibody, clone TLD-3A12, azide free detects level of PECAM-1 and has been published and validated for use in ELISA, flow cytometry, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PECAM-1
Applications ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone TLD-3A12
Conjugate Unconjugated
Formulation Purified immunoglobulin by affinity chromatography on Protein G. Liquid in phosphate buffered saline containing no preservatives.
Gene Symbols ENSG00000188536, HBA2, Alpha-globin
Host Species Mouse
Immunogen Activated, Lewis rat derived microglial cells.
Quantity 100 μg
Primary or Secondary Primary
Target Species Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.