Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PECAM-1 Rat anti-Mouse, FITC, Clone: 390, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma CBL1337F
Description
This Anti-PECAM-1 Antibody, clone 390, FITC conjugated is validated for use in flow cytometry, immunohistochemistry for the detection of PECAM-1.
Specifications
PECAM-1 | |
Monoclonal | |
FITC | |
NM_000442.3 | |
Rat | |
500 μg | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry | |
390 | |
Purified immunoglobulin conjugated to fluorescein. Liquid in in phosphate buffered saline containing 0.1% sodium azide. | |
PECAM1, EndoCAM, GPIIA' , PECAM-1, CD31, CD31/EndoCAM | |
Mouse leukocyte cell line 32D | |
Primary | |
2-8°C for up to 12 months. Do not Freeze. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction