Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PEMT Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA511145
Description
PEMT is an enzyme which converts phosphatidylethanolamine to phosphatidylcholine by sequential methylation in the liver. The protein localizes to the endoplasmic reticulum and mitochondria-associated membranes. The gene is within the Smith-Magenis syndrome region on chromosome 17.
Specifications
PEMT | |
Polyclonal | |
Unconjugated | |
Q9UBM1 | |
PEMT | |
KLH conjugated synthetic peptide between 170-199 amino acids from the C-terminal region of human PEMT | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
1.5 mg/mL | |
PEMT | |
PEAMT; PEMT2 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
10400 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction