Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Peptidyl-citrulline Mouse anti-Human, Mouse, Chicken, Rat, Clone: F95

Catalog No. MABN328MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN328MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN328MI Supplier MilliporeSigma™ Supplier No. MABN328
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-peptidyl-citrulline, clone F95 Antibody is validated for use in Immunohistochemistry and Western Blotting and Immunocytochemistry for the detection of peptidyl-citrulline.
TRUSTED_SUSTAINABILITY

Specifications

Antigen peptidyl-citrulline
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone F95
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
Host Species Mouse
Immunogen Citrullinated peptide consisting of 10 citrulline residues and a carboxyl Gly-Gly-Cys.
Purification Method Purified by ion-exchange chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Chicken, Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.