Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perforin Mouse anti-Human, Alexa Fluor 647, Clone: deltaG9, BD

Catalog No. BDB563576 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563576 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563576 Supplier BD Biosciences Supplier No. 563576
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Perforin has a key role in cell-mediated cytotoxicity. It is a 70 kDa cytolytic protein that is expressed in the cytoplasmic granules of cytotoxic T lymphocytes (CTLs) and natural killer (NK) cells. CTLs are involved in eliminating virally infected cells, in anti-tumor immune responses, in allograft rejections, and in some autoimmune diseases. NK cells are important for tumor surveillance and destruction and are involved in allograft rejections. Cytotoxic cells release the contents of their cytotoxic granules, including perforin upon recognition of their target cell. In the presence of calcium, perforin forms transmembrane channels or pores in the membrane of the target cell leading to a cell death that resembles apoptosis. The ability to detect perforin-positive cells with specific antibody should be useful in identifying and understanding perforin-mediated reactions.

Clone δG9 reacts with human and bovine perforin. It does not cross-react with mouse perforin. Purified granules from the human lymphoma cell line YT were used as immunogen. Clone δG9 was initially characterized by immunoprecipitation and immunohistochemistry of frozen tissue sections. The antibody stains scattered lymphocytes in red pulp of spleen, and scattered infiltrated lymphocytes in lymphoma.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Perforin
Applications Flow Cytometry
Classification Monoclonal
Clone deltaG9
Conjugate Alexa Fluor 647
Description PRF1; P1; PERF; PFN1; PFP; Perforin-1; Cytolysin; FLH2; HPLH2
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Purified Granules from the Human Lymphoma Cell Line YT
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bovine, Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.