Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perforin Mouse anti-Human, PE, Clone: ΔG9, BD

Catalog No. BDB556437 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556437 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556437 Supplier BD Biosciences Supplier No. 556437
Add to Cart
Add to Cart

Includes

PE Mouse Anti-Human Perforin, PE Mouse IgG2b, κ Isotype Control (anti-dansyl)

Mouse Monoclonal Antibody

Perforin has a key role in cell-mediated cytotoxicity. It is a 70kDa cytolytic protein that is expressed in the cytoplasmic granules of cytotoxic T lymphocytes (CTLs) and natural killer (NK) cells. CTLs are involved in eliminating virally infected cells, in anti-tumor immune responses, in allograft rejections, and in some autoimmune diseases. Cytotoxic cells release the contents of their cytotoxic granules, including perforin upon recognition of their target cell. In the presence of calcium, perforin forms transmembrane channels or pores in the membrane of the target cell leading to a cell death that resembles apoptosis. The ability to detect perforin-positive cells with specific antibody should be useful in identifying and understanding perforin-mediated reactions. Clone δG9 reacts with human and bovine perforin. It does not cross-react with mouse perforin. Clone δG9 was initially characterized by immunoprecipitation and immunohistochemistry of frozen tissue sections, and has also been published for flow cytometric analysis. Clone 27-35 is an isotype control monoclonal antibody specific for hapten dansyl (5-[dimethylamino] naphthalene-1-sulfonyl). This hapten is not expressed on human cells or human cell lines.

Host Species: Mouse
Clone: δG9, 27-35
Isotype: IgG2b, κ
Species Reactivity: Human
Immunogen: Purified Granules from the Human Lymphoma Cell Line YT

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Perforin
Applications Flow Cytometry
Classification Monoclonal
Clone ΔG9
Conjugate PE
Host Species Mouse
Immunogen Purified Granules from the Human Lymphoma Cell Line YT
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.