Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pericentrin Mouse anti-Mouse, Unlabeled, Clone: 30, BD

Catalog No. BDB611814 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611814 50 μg
BDB611815 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611814 Supplier BD Biosciences Supplier No. 611814
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Centrosomes and other microtubule-organizing centers are a diverse group of organelles that nucleate and organize microtubules for many celllular processes, such as mitotic spindle formation, organelle tranpsort, and protein localization. Pericentrin is a protein in the pericentriolar material, a filamentous matrix surrounding centrioles, which can organize microtubule spindle formation. The structure of pericentrin includes multiple α-helical domains that form coiled-coil domains separated by non-helical, non-coiled regions. It is found in a 3MDa-complex that includes γ-tubulin, a form of tubulin that nucleates microtubule formation at the centrosome. Pericentrin exhibits the highest expression in embryonic mouse kidney, thymus, and liver. Injection of pericentrin antibodies in Xenopus oocytes disrupts mitotic and meiotic divisions and blocks microtubule aster formation. In addition, dynein transports both pericentrin and γ-tubulin to centrosomes along microtubules in Xenopus oocyte extracts. Thus, the complex of pericentrin and γ-tubulin may be recruited to the centrosome by dynein, where the complex becomes anchored to the centrosome for microtubule nucleating activity.

Host Species: Mouse
Clone: 30
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Mouse pericentrin aa. 1692-1814

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Pericentrin
Applications Western Blot
Classification Monoclonal
Clone 30
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse pericentrin aa. 1692-1814
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.